siRNA Details
Gene Symbol: MIR194-2HG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_133638.1 | 2 | 1265 | |||
GenBank mRNA | AK092802.1 | - | 2 | 1265 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: MIR194-2HG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_133638.1 | 2 | 1265 | |||
GenBank mRNA | AK092802.1 | - | 2 | 1265 |