siRNA Details
| Gene Symbol: MIR194-2HG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_133638.1 | 2 | 626 | |||
| NR_133639.1 | 2 | 285 | ||||
| GenBank mRNA | AB429223.1 | - | 2 | 285 | ||
| AK092802.1 | - | 2 | 626 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MIR194-2HG| Gene Symbol: MIR194-2HG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_133638.1 | 2 | 626 | |||
| NR_133639.1 | 2 | 285 | ||||
| GenBank mRNA | AB429223.1 | - | 2 | 285 | ||
| AK092802.1 | - | 2 | 626 | |||