siRNA Details
| Gene Symbol: GPR1-AS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104359.1 | 6 | 2446 | |||
| GenBank mRNA | AK002005.1 | - | 1 | 918 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

GPR1-AS| Gene Symbol: GPR1-AS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104359.1 | 6 | 2446 | |||
| GenBank mRNA | AK002005.1 | - | 1 | 918 | ||