siRNA Details
Gene Symbol: GPR1-AS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_104359.1 | 6 | 3011 | |||
GenBank mRNA | AK002005.1 | - | 1 | 1483 | ||
CB160399.1 | - | 1 | 292 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: GPR1-AS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_104359.1 | 6 | 3011 | |||
GenBank mRNA | AK002005.1 | - | 1 | 1483 | ||
CB160399.1 | - | 1 | 292 |