siRNA Details
| Gene Symbol: NPAS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK002000.1 | - | 1 | 1673 | ||
| AK021667.1 | - | 1 | 914 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

NPAS3| Gene Symbol: NPAS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK002000.1 | - | 1 | 1673 | ||
| AK021667.1 | - | 1 | 914 | |||