siRNA Details
| Gene Symbol: LOC101927795 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110275.1 | 2, 3 | 471 | |||
| GenBank mRNA | AY245869.1 | - | 3, 4 | 459 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CR748201.1 | - | 3, 4 | 435 | Chr. 2 - 200791482 - 200812170 [-] on Build GRCh38 | |

LOC101927795
Genomic Map