siRNA Details
Gene Symbol: LOC101927795 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110275.1 | 4 | 766 | |||
GenBank mRNA | AY245869.1 | - | 5 | 754 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC101927795 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110275.1 | 4 | 766 | |||
GenBank mRNA | AY245869.1 | - | 5 | 754 |