siRNA Details
| Gene Symbol: BCL6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005247694.3 | XP_005247751.1 | 1 | 2544 | ||
| GenBank mRNA | AK090890.1 | - | 1 | 1226 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

BCL6| Gene Symbol: BCL6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005247694.3 | XP_005247751.1 | 1 | 2544 | ||
| GenBank mRNA | AK090890.1 | - | 1 | 1226 | ||