siRNA Details
| Gene Symbol: BCL6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005247694.3 | XP_005247751.1 | 1 | 2038 | ||
| GenBank mRNA | AK090890.1 | - | 1 | 720 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BQ883719.1 | - | 1 | 22 | Chr. 3 - 187738341 - 187739240 [-] on Build GRCh38 | |

BCL6
Genomic Map