siRNA Details
| Gene Symbol: KATNBL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK311338.1 | - | 6 | 1511 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK021466.1 | - | 1 | 1221 | Chr. 15 - 34145967 - 34147487 [-] on Build GRCh38 | |

KATNBL1
Genomic Map