siRNA Details
| Gene Symbol: LOC284630 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK096384.1 | - | 1 | 1408 | ||
| Gene Symbol: TTC34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_016999990.1 | XP_016855479.1 | 9 | 4329 | ||

LOC284630
Genomic Map