siRNA Details
| Gene Symbol: IMPDH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | J05272.1 | - | 1 | 21 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DQ067456.1 | - | 1 | 216 | Chr. 7 - 128410218 - 128410447 [-] on Build GRCh38 | |

IMPDH1
Genomic Map