siRNA Details
| Gene Symbol: LINC00642 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033986.1 | 3 | 292 | |||
| GenBank mRNA | AK093266.1 | - | 2 | 1610 | ||
| BC036259.1 | - | 2 | 959 | |||
| BX101323.1 | - | 3 | 292 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00642| Gene Symbol: LINC00642 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033986.1 | 3 | 292 | |||
| GenBank mRNA | AK093266.1 | - | 2 | 1610 | ||
| BC036259.1 | - | 2 | 959 | |||
| BX101323.1 | - | 3 | 292 | |||