siRNA Details
| Gene Symbol: LINC00664 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037194.1 | 6 | 1133 | |||
| GenBank mRNA | AK097381.1 | - | 11 | 1540 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00664| Gene Symbol: LINC00664 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037194.1 | 6 | 1133 | |||
| GenBank mRNA | AK097381.1 | - | 11 | 1540 | ||