siRNA Details
| Gene Symbol: LINC00664 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037194.1 | 8 | 1492 | |||
| GenBank mRNA | AK097381.1 | - | 13 | 1899 | ||
| BX537835.1 | - | 8 | 1492 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DB039272.1 | - | 2 | 227 | Chr. 19 - 21494141 - 21498758 [+] on Build GRCh38 | |

LINC00664
Genomic Map