siRNA Details
| Gene Symbol: GPATCH2L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006720191.2 | XP_006720254.1 | 10 | 13145 | ||
| XM_017021427.1 | XP_016876916.1 | 11 | 13387 | |||
| XM_017021428.1 | XP_016876917.1 | 11 | 13372 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

GPATCH2L| Gene Symbol: GPATCH2L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006720191.2 | XP_006720254.1 | 10 | 13145 | ||
| XM_017021427.1 | XP_016876916.1 | 11 | 13387 | |||
| XM_017021428.1 | XP_016876917.1 | 11 | 13372 | |||