siRNA Details
| Gene Symbol: PKIA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006823.3 | NP_006814.1 | 4 | 3970 | ||
| NM_181839.2 | NP_862822.1 | 3 | 3841 | |||
| XM_017013620.1 | XP_016869109.1 | 5 | 3845 | |||
| XM_017013621.1 | XP_016869110.1 | 4 | 3811 | |||
| XM_017013622.1 | XP_016869111.1 | 3 | 3576 | |||
| GenBank mRNA | CR744438.1 | - | 1 | 371 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CR595807.1 | - | 1 | 1019 | Chr. 8 - 78604018 - 78605266 [+] on Build GRCh38 | |

PKIA
Genomic Map