siRNA Details
| Gene Symbol: GPATCH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017001593.1 | XP_016857082.1 | 9 | 2286 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

GPATCH2| Gene Symbol: GPATCH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017001593.1 | XP_016857082.1 | 9 | 2286 | ||