siRNA Details
Gene Symbol: FBRSL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005266181.3 | XP_005266238.1 | 3 | 6566 | ||
GenBank mRNA | BC008052.2 | - | 1 | 1324 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: FBRSL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005266181.3 | XP_005266238.1 | 3 | 6566 | ||
GenBank mRNA | BC008052.2 | - | 1 | 1324 |