siRNA Details
| Gene Symbol: LOC284108 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK093715.1 | - | 1 | 1589 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CR616125.1 | - | 1 | 1577 | Chr. 17 - 39165085 - 39167849 [+] on Build GRCh38 | |

LOC284108
Genomic Map