siRNA Details
| Gene Symbol: LOC84843 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC007304.1 | - | 1 | 575 | 3352097 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC84843| Gene Symbol: LOC84843 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC007304.1 | - | 1 | 575 | 3352097 | |