siRNA Details
Gene Symbol: LOC100508631 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC042993.1 | - | 2 | 637 | ||
BG621125.1 | - | 2 | 668 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC100508631 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC042993.1 | - | 2 | 637 | ||
BG621125.1 | - | 2 | 668 |