siRNA Details
| Gene Symbol: LOC100506459 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK126833.1 | - | 2 | 742 | ||
| AL832882.1 | - | 1 | 2834 | |||
| Gene Symbol: CD58 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017002869.1 | XP_016858358.1 | 6 | 11428 | ||

LOC100506459
Genomic Map