siRNA Details
| Gene Symbol: DHODH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005255827.3 | XP_005255884.1 | 9 | 3385 | ||
| XM_005255829.3 | XP_005255886.1 | 8 | 3022 | |||
| XM_017022990.1 | XP_016878479.1 | 9 | 3160 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK098239.1 | - | 2 | 719 | Chr. 16 - 72025041 - 72027663 [+] on Build GRCh38 | |

DHODH
Genomic Map