siRNA Details
| Gene Symbol: LINC01070 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_108094.1 | 3 | 338 | |||
| GenBank mRNA | CR627049.1 | - | 2 | 1879 | ||
| HY055056.1 | - | 3 | 338 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01070| Gene Symbol: LINC01070 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_108094.1 | 3 | 338 | |||
| GenBank mRNA | CR627049.1 | - | 2 | 1879 | ||
| HY055056.1 | - | 3 | 338 | |||