siRNA Details
| Gene Symbol: NEK4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006713310.2 | XP_006713373.1 | 15 | 5660 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK022015.1 | - | 1518 | Chr. - - [-] on Build | ||

NEK4
Genomic Map