siRNA Details
Gene Symbol: LOC101929297 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125862.1 | 3, 4 | 1299 | |||
GenBank mRNA | BC040873.1 | - | 3, 4 | 1299 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC101929297 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125862.1 | 3, 4 | 1299 | |||
GenBank mRNA | BC040873.1 | - | 3, 4 | 1299 |