siRNA Details
Gene Symbol: LOC101929297 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125862.1 | 1, 2 | 75 | |||
GenBank mRNA | BC040873.1 | - | 1, 2 | 75 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BM906286.1 | - | 1, 2 | 94 | Chr. 6 - 166236967 - 166243625 [+] on Build GRCh38 |
