siRNA Details
Gene Symbol: KCND3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005270851.4 | XP_005270908.1 | 8 | 6706 | ||
XM_006710629.3 | XP_006710692.1 | 8 | 7016 | |||
XM_006710630.3 | XP_006710693.1 | 7 | 6649 | |||
XM_017001244.1 | XP_016856733.1 | 8 | 7031 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AF070632.1 | - | 2 | 587 | Chr. 1 - 111770662 - 111772569 [-] on Build GRCh38 |
