siRNA Details
| Gene Symbol: KCND3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005270851.4 | XP_005270908.1 | 8 | 7293 | ||
| XM_006710629.3 | XP_006710692.1 | 8 | 7603 | |||
| XM_006710630.3 | XP_006710693.1 | 7 | 7236 | |||
| XM_017001244.1 | XP_016856733.1 | 8 | 7618 | |||

KCND3
Genomic Map