siRNA Details
| Gene Symbol: LOC101927877 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135634.1 | 1 | 251 | |||
| GenBank mRNA | BC040294.1 | - | 1 | 253 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BG717414.1 | - | 1 | 258 | Chr. 17 - 62808496 - 62835340 [+] on Build GRCh38 | |

LOC101927877
Genomic Map