siRNA Details
| Gene Symbol: LOC101927877 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135634.1 | 2 | 1705 | |||
| GenBank mRNA | BC040294.1 | - | 2 | 1706 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000584542.1 | - | 2 | 1707 | Chr. 17 - 62808500 - 62836371 [+] on Build GRCh38 | |

LOC101927877
Genomic Map