siRNA Details
| Gene Symbol: LOC100131540 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK129921.1 | - | 1 | 1070 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100131540| Gene Symbol: LOC100131540 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK129921.1 | - | 1 | 1070 | ||