siRNA Details
| Gene Symbol: LOC100506444 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125918.1 | 3 | 737 | |||
| GenBank mRNA | AK129929.1 | - | 3 | 737 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BU189569.1 | - | 3 | 749 | Chr. 4 - 53695915 - 53733585 [+] on Build GRCh38 | |

LOC100506444
Genomic Map