siRNA Details
| Gene Symbol: LOC100506444 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125918.1 | 3 | 1382 | |||
| GenBank mRNA | AK129929.1 | - | 3 | 1382 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100506444| Gene Symbol: LOC100506444 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125918.1 | 3 | 1382 | |||
| GenBank mRNA | AK129929.1 | - | 3 | 1382 | ||