siRNA Details
Gene Symbol: LOC100506444 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125918.1 | 3 | 465 | |||
GenBank mRNA | AK129929.1 | - | 3 | 465 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BU189569.1 | - | 3 | 474 | Chr. 4 - 53695915 - 53733585 [+] on Build GRCh38 |
