siRNA Details
| Gene Symbol: LOC100506444 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125918.1 | 3 | 1240 | |||
| GenBank mRNA | AK129929.1 | - | 3 | 1240 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000506950.1 | - | 3 | 1240 | Chr. 4 - 53695901 - 53734358 [+] on Build GRCh38 | |

LOC100506444
Genomic Map