siRNA Details
| Gene Symbol: LOC100132071 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK128253.1 | - | 1 | 2860 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100132071| Gene Symbol: LOC100132071 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK128253.1 | - | 1 | 2860 | ||