siRNA Details
| Gene Symbol: MEG3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046473.1 | 7 | 4380 | |||
| GenBank mRNA | AK098378.1 | - | 1 | 1329 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MEG3| Gene Symbol: MEG3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046473.1 | 7 | 4380 | |||
| GenBank mRNA | AK098378.1 | - | 1 | 1329 | ||