siRNA Details
Gene Symbol: HFE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011514543.2 | XP_011512845.1 | 7 | 2497 | ||
GenBank mRNA | AF147392.1 | - | 1 | 203 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: HFE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011514543.2 | XP_011512845.1 | 7 | 2497 | ||
GenBank mRNA | AF147392.1 | - | 1 | 203 |