siRNA Details
| Gene Symbol: UBE2G2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC054341.1 | - | 1 | 551 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BQ424505.1 | - | 1 | 559 | Chr. 21 - 44801004 - 44801794 [-] on Build GRCh38 | |

UBE2G2
Genomic Map