siRNA Details
| Gene Symbol: PCF11-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_133914.1 | 1 | 671 | |||
| GenBank mRNA | AK092827.1 | - | 1 | 223 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DA925269.1 | - | 1 | 224 | Chr. 11 - 83186445 - 83187036 [-] on Build GRCh38 | |

PCF11-AS1
Genomic Map