siRNA Details
Gene Symbol: GRTP1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046541.1 | 2 | 1226 | |||
GenBank mRNA | BC037976.1 | - | 2 | 1222 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: GRTP1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046541.1 | 2 | 1226 | |||
GenBank mRNA | BC037976.1 | - | 2 | 1222 |