siRNA Details
| Gene Symbol: RNPS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005255049.3 | XP_005255106.1 | 1 | 1100 | ||
| GenBank mRNA | AK021799.1 | - | 1 | 403 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RNPS1| Gene Symbol: RNPS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005255049.3 | XP_005255106.1 | 1 | 1100 | ||
| GenBank mRNA | AK021799.1 | - | 1 | 403 | ||