siRNA Details
| Gene Symbol: STK32C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001318878.1 | NP_001305807.1 | 1 | 387 | ||
| NM_001318880.1 | NP_001305809.1 | 1 | 387 | |||
| NM_001318881.1 | NP_001305810.1 | 1 | 387 | |||
| NM_001318882.1 | NP_001305811.1 | 1 | 387 | |||
| XM_011539688.1 | XP_011537990.1 | 1 | 405 | |||
| XM_017016100.1 | XP_016871589.1 | 1 | 405 | |||
| XM_017016101.1 | XP_016871590.1 | 1 | 405 | |||
| GenBank mRNA | AK302102.1 | - | 1 | 334 | ||
| AK308956.1 | - | 1 | 365 | |||
| BC042566.1 | - | 1 | 364 | |||
| CD674243.1 | - | 1 | 381 | |||
| DA574949.1 | - | 1 | 364 | |||
| HY020713.1 | - | 1 | 387 | |||
| HY070240.1 | - | 1 | 357 | |||
| HY106710.1 | - | 1 | 362 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DN997246.1 | - | 1 | 356 | Chr. 10 - 132330294 - 132331837 [-] on Build GRCh38 | |

STK32C
Genomic Map