siRNA Details
| Gene Symbol: FBXL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005266930.2 | XP_005266987.1 | 9 | 6821 | ||
| XM_017010727.1 | XP_016866216.1 | 8 | 6675 | |||
| XM_017010728.1 | XP_016866217.1 | 11 | 6981 | |||
| GenBank mRNA | AK055220.1 | - | 1 | 1441 | ||

FBXL4
Genomic Map