siRNA Details
| Gene Symbol: NEMF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017021760.1 | XP_016877249.1 | 32 | 5263 | ||
| XM_017021761.1 | XP_016877250.1 | 27 | 4716 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

NEMF| Gene Symbol: NEMF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017021760.1 | XP_016877249.1 | 32 | 5263 | ||
| XM_017021761.1 | XP_016877250.1 | 27 | 4716 | |||