siRNA Details
| Gene Symbol: PEX11A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271572.1 | NP_001258501.1 | 3 | 2604 | ||
| NM_001271573.1 | NP_001258502.1 | 2 | 2581 | |||
| NM_003847.2 | NP_003838.1 | 3 | 2697 | |||
| GenBank mRNA | AL109728.1 | - | 1 | 809 | ||
| AL360141.1 | - | 1 | 1811 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BQ437764.1 | - | 1 | 587 | Chr. 15 - 89681535 - 89682162 [-] on Build GRCh38 | |

PEX11A
Genomic Map