siRNA Details
| Gene Symbol: PEX11A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271572.1 | NP_001258501.1 | 3 | 1467 | ||
| NM_001271573.1 | NP_001258502.1 | 2 | 1444 | |||
| NM_003847.2 | NP_003838.1 | 3 | 1560 | |||
| GenBank mRNA | AK297442.1 | - | 2 | 1292 | ||
| AL360141.1 | - | 1 | 674 | |||

PEX11A
Genomic Map