siRNA Details
| Gene Symbol: DERL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001304777.1 | NP_001291706.1 | 7 | 4071 | ||
| NM_001304779.1 | NP_001291708.1 | 4 | 3710 | |||
| NM_016041.4 | NP_057125.2 | 7 | 4074 | |||
| NR_130905.1 | 6 | 4186 | ||||
| NR_130906.1 | 6 | 3980 | ||||
| GenBank mRNA | AK092218.1 | - | 1 | 1840 | ||

DERL2
Genomic Map